Methyl salicylate ≥98%

Supplier: BeanTown Chemical

Synonyms: Wintergreen Oil, Methyl 2-hydroxybenzoate

217500-2.5KG 217500-500G
BT217500-2.5KGEA 137.72 USD
BT217500-2.5KG BT217500-500G
Methyl salicylate ≥98%
Methyl salicylate

CAS: 119-36-8; EC No: 204-317-7; MDL No: MFCD00002214; RTECS: VO4725000 Liquid; Linear Formula: HOC6H4COOCH3; Molecular Formula: C8H8O3; MW: 152.15 Melting Point: -8°; Boiling Point: 222°; Flash point: 96°C (205°F) Density (g/mL): 1.174; Refractive Index: 1.536 Light Sensitive


  • RTECS: VO4725000
  • Refractive index: 1.536
  • Light Sensitive
  • Liquid

Formula: 2-(HO)C₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 432
Melting Pt: –8 °C
Density: 1.174 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR