Ammonium molybdate(VI) ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Molybdic acid ammonium salt, Molybdic acid, ammonium salt, Ammonium molybdate, di-Ammonium molybdate

217410-100G 217410-25G
BT217410-100GEA 629.68 USD
BT217410-100G BT217410-25G
Ammonium molybdate(VI) ≥99.99%
Ammonium molybdate(VI)

CAS: 13106-76-8; EC No: 236-031-3; MDL No: MFCD00011596; RTECS: QA4900000 Powder/Lump; Linear Formula: (NH4)2MoO4; MW: 196.01


  • RTECS: QA4900000
  • Powder/Lump

Formula: H₈MoN₂O₄
MDL Number: MFCD00011596
CAS Number: 13106-76-8
EINECS: 236-031-3

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR