Magnesium oxide ≥95%

Supplier: BeanTown Chemical

Synonyms: Magnesia usta

217240-100G 217240-2KG 217240-500G
BT217240-100GEA 81.93 USD
BT217240-100G BT217240-2KG BT217240-500G
Magnesium oxide ≥95%
Magnesium oxide

CAS: 1309-48-4; EC No: 215-171-9; MDL No: MFCD00011109 -140+325 Mesh Powder; Molecular Formula: MgO ; MW: 40.30 Melting Point: 2852°; Boiling Point: 3600° Density (g/mL): 3.58; Refractive Index: 1.736 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.736
  • Air Sensitive, Moisture Sensitive
  • -140+325 Mesh Powder

Formula: MgO
Boiling Pt: 3600 °C
Melting Pt: 2852 °C
Density: 3.58 g/mL
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR