Polyethylene glycol 20,000
Supplier: BeanTown Chemical
Synonyms:
PEG 20000
, PEG 20,000
, Ethylene glycol polymer 20000
217205-1KG
217205-250G
BT217205-250GEA
45.92
USD
BT217205-250G
BT217205-1KG
Polyethylene glycol 20,000
Polyethylene glycol 20,000
- Flakes
Formula:
H(OCH₂CH₂)nOH MW: 20000 g/mol Melting Pt: 63…66 °C Density: 1.2 g/mL Flash Pt: 229 °C (444 °F) |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 203-473-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...