Magnesium acetate tetrahydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

216995-1KG 216995-250G
BT216995-1KGEA 91.04 USD
BT216995-1KG BT216995-250G
Magnesium acetate tetrahydrate ≥98%
Magnesium acetate tetrahydrate

CAS: 16674-78-5; EC No: 205-554-9; MDL No: MFCD00149214 Crystalline; Linear Formula: (CH3COO)2Mg·4H2O; MW: 214.45 Melting Point: 72-75° Density (g/mL): 1.454 Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 72…75 °C
Density: 1.454 g/mL
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR