Dimethyl methoxymethylenemalonate ≥98%

Supplier: BeanTown Chemical

216925-10G 216925-250G 216925-50G
BT216925-10GEA 41 USD
BT216925-10G BT216925-250G BT216925-50G
Dimethyl methoxymethylenemalonate ≥98%
Dimethyl methoxymethylenemalonate

CAS: 22398-14-7; EC No: 244-957-4; MDL No: MFCD00038225 Crystalline; Molecular Formula: C7H10O5; MW: 174.16 Melting Point: 41-45°; Boiling Point: 97-99°/0.5 mmHg


  • Crystalline

Formula: C₇H₁₀O₅
Boiling Pt: 97…99 °C/0.5 mmHg
Melting Pt: 41…45 °C
MDL Number: MFCD00038225
CAS Number: 22398-14-7
EINECS: 244-957-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR