4-Ethoxy-3-nitropyridine hydrochloride ≥97%

Supplier: BeanTown Chemical

216870-25G 216870-5G
BT216870-25GEA 183 USD
BT216870-25G BT216870-5G
4-Ethoxy-3-nitropyridine hydrochloride ≥97%
4-Ethoxy-3-nitropyridine hydrochloride

CAS: 94602-04-7; MDL No: MFCD01075663 Powder; Molecular Formula: C7H9ClN2O3; MW: 204.61 Melting Point: 270-272° Hygroscopic


  • Hygroscopic
  • Powder

Formula: C₇H₉ClN₂O₃
Melting Pt: 270…272 °C
MDL Number: MFCD01075663
CAS Number: 94602-04-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR