Rhodamine B

Supplier: BeanTown Chemical

216735-1KG 216735-250G 216735-50G
BT216735-1KGEA 225 USD
BT216735-1KG BT216735-250G BT216735-50G
Rhodamine B
Rhodamine B

CAS: 81-88-9; EC No: 201-383-9; MDL No: MFCD00011931; RTECS: BP3675000 Powder; Molecular Formula: C28H31ClN2O3; MW: 479.02 Melting Point: 210-211° (decomposes) Density (g/mL): 0.790


  • RTECS: BP3675000
  • Powder

Formula: C₂₈H₃₁ClN₂O₃
Melting Pt: 210…211 °C (dec.)
Density: 0.790 g/mL
MDL Number: MFCD00011931
CAS Number: 81-88-9
EINECS: 201-383-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR