Diethyl (2-oxopropyl)phosphonate ≥97%

Supplier: BeanTown Chemical

Synonyms: Diethyl acetylmethylphosphonate

216665-1G 216665-25G 216665-5G
BT216665-1GEA 40 USD
BT216665-1G BT216665-25G BT216665-5G
Diethyl (2-oxopropyl)phosphonate ≥97%
Diethyl (2-oxopropyl)phosphonate

CAS: 1067-71-6; MDL No: MFCD00044728 Liquid; Molecular Formula: C7H15O4P; MW: 194.17 Boiling Point: 126°/99 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.01; Refractive Index: 1.433


  • Refractive index: 1.433
  • Liquid

Formula: C₇H₁₅O₄P
MW: 194.17 g/mol
Boiling Pt: 126 °C/99 mmHg
Density: 1.01 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00044728
CAS Number: 1067-71-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR