Silica gel, Pore size: 60 Å 0.060-0.2mm; 70-230 mesh
Supplier: BeanTown Chemical
216420-1KG
216420-5KG
BT216420-1KGEA
127.77
USD
BT216420-1KG
BT216420-5KG
Silica gel, Pore size: 60 Å 0.060-0.2mm; 70-230 mesh
Silica gel
CAS: 63231-67-4; EC No: 231-545-4; MDL No: MFCD00011232; RTECS: VV7565000
Powder
Melting Point:
- RTECS: VV7565000
- Hygroscopic
- Powder
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: >1600 °C |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...