Tungsten ≥99.95% (trace metals basis), wire Ø 0.75 mm
Supplier: BeanTown Chemical
216365-1M
216365-5M
BT216365-1MEA
54
USD
BT216365-1M
BT216365-5M
Tungsten ≥99.95% (trace metals basis), wire Ø 0.75 mm
Tungsten
CAS: 7440-33-7; EC No: 231-143-9; MDL No: MFCD00011461
Solid; Molecular Formula: W; MW: 183.84
Melting Point: 3410°; Boiling Point: 5660°
Density (g/mL): 19.3
- Solid
Formula:
W Boiling Pt: 5660 °C Melting Pt: 3410 °C Density: 19.3 g/mL |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...