Hydrochloric acid 0.01 N in water volumetric solution
Supplier: BeanTown Chemical
Synonyms:
Hydrochloric acid solution, Muriatic acid
216250-1L
216250-20L
216250-4L
BT216250-1LEA
31.32
USD
BT216250-1L
BT216250-20L
BT216250-4L
Hydrochloric acid 0.01 N in water volumetric solution
Hydrochloric acid
CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324; RTECS: MW4025000
Liquid; Molecular Formula: HCl; MW: 36.46
- RTECS: MW4025000
- Liquid
Formula:
HCl |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...