Magnesium fluoride ≥99% (trace metals basis excluding Ca, Na)

Supplier: BeanTown Chemical

216030-1KG 216030-250G 216030-50G
BT216030-1KGEA 815 USD
BT216030-1KG BT216030-250G BT216030-50G
Magnesium fluoride ≥99% (trace metals basis excluding Ca, Na)
Magnesium fluoride

CAS: 7783-40-6; EC No: 231-995-1; MDL No: MFCD00011108; RTECS: OM3325000 -200 Mesh Powder; Linear Formula: MgF2; MW: 62.30 Melting Point: 1621° Density (g/mL): 3.148


  • RTECS: OM3325000
  • -200 Mesh Powder

Formula: MgF₂
MW: 62.3 g/mol
Boiling Pt: 2260
Melting Pt: 1266
Density: 3.148 g/mL
MDL Number: MFCD00011108
CAS Number: 7783-40-6
EINECS: 231-995-1
Merck Index: 12,05701

Order Now

Specification Test Results

Assay 99%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR