4-Aminobenzoic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1
215950-1KG
215950-250G
215950-50G
BT215950-1KGEA
120
USD
BT215950-1KG
BT215950-250G
BT215950-50G
4-Aminobenzoic acid ≥99%
4-Aminobenzoic acid
CAS: 150-13-0; EC No: 205-753-0; MDL No: MFCD00007894; RTECS: DG1400000
Solid; Molecular Formula: C7H7NO2; MW: 137.14
Melting Point: 187-189°
Density (g/mL): 1.374
Air Sensitive, Light Sensitive
- RTECS: DG1400000
- Air Sensitive, Light Sensitive
- Solid
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 340 Melting Pt: 188 Density: 1.374 g/mL |
MDL Number:
MFCD00007894 CAS Number: 150-13-0 EINECS: 205-753-0 Merck Index: 13,00422 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...