Vinylferrocene
Supplier: BeanTown Chemical
215840-1G
215840-5G
BT215840-1GEA
121.51
USD
BT215840-1G
BT215840-5G
Vinylferrocene
Vinylferrocene
- Crystalline
Formula:
C₁₂H₁₂Fe Boiling Pt: 80…85 °C/0.2 mmHg Melting Pt: 51…53 °C Flash Pt: 62 °C (144 °F) |
MDL Number:
MFCD00001434 EINECS: 215-041-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...