Potassium bromide 99,0 ACS

Supplier: BeanTown Chemical

215760-1KG 215760-250G 215760-50G
BT215760-1KGEA 138.07 USD
BT215760-1KG BT215760-250G BT215760-50G
Potassium bromide 99,0 ACS
Potassium bromide

CAS: 7758-02-3; EC No: 231-830-3; MDL No: MFCD00011358; RTECS: TS7650000 Crystalline; Linear Formula: KBr; MW: 119.00 Melting Point: 734°; Boiling Point: 1435° Density (g/mL): 2.75; Refractive Index: 1.559 Hygroscopic


  • RTECS: TS7650000
  • Refractive index: 1.559
  • Hygroscopic
  • Crystalline

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75 g/mL
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR