Sodium chloride 99
Supplier: BeanTown Chemical
Synonyms:
Saline, NaCl, Sea salt, Halite
215700-1KG
215700-5KG
BT215700-1KGEA
33.98
USD
BT215700-1KG
BT215700-5KG
Sodium chloride 99
Sodium chloride
CAS: 7647-14-5; EC No: 231-598-3; MDL No: MFCD00003477; RTECS: VZ4725000
Powder; Linear Formula: NaCl; MW: 58.44
Melting Point: 801°; Boiling Point: 1413°
Density (g/mL): 2.165; Refractive Index: 1.544
- RTECS: VZ4725000
- Refractive index: 1.544
- Powder
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 Melting Pt: 801 Density: 2.165 g/mL |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...