Sodium trifluoromethanesulfonate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Trifluoromethanesulphonic acid sodium salt, Trifluoromethanesulfonic acid sodium salt
215675-100G
215675-25G
215675-5G
BT215675-100GEA
466
USD
BT215675-100G
BT215675-25G
BT215675-5G
Sodium trifluoromethanesulfonate ≥98%
Sodium trifluoromethanesulphonate
CAS: 2926-30-9; MDL No: MFCD00061607
Crystalline; Linear Formula: CF2SO3Na; MW: 172.06
Melting Point: 253-255°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
CF₃NaO₃S MW: 172.06 g/mol Melting Pt: 253…255 °C |
MDL Number:
MFCD00061607 CAS Number: 2926-30-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...