Platinum rhodium alloy (90:10 w%) ≥99.9% (trace metals basis), foil, Thickness 0.25 mm

Supplier: BeanTown Chemical

215475-100X100 215475-25X25MM 215475-50X50MM
BT215475-100X1EA 1216 USD
BT215475-100X1 BT215475-25X25 BT215475-50X50
Platinum rhodium alloy (90:10 w%) ≥99.9% (trace metals basis), foil, Thickness 0.25 mm
Platinum Rhodium alloy (90:10 w%)

MDL No: MFCD00198186 Solid; Formula: Pt:Rh; 90:10 wt%


  • Solid

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR