Guar gum

Supplier: BeanTown Chemical

215250-1KG 215250-250G 215250-500G
BT215250-1KGEA 179.01 USD
BT215250-1KG BT215250-250G BT215250-500G
Guar gum
Guar gum

CAS: 9000-30-0; EC No: 232-536-8; MDL No: MFCD00131250 Powder


  • Powder

MDL Number: MFCD00131250
CAS Number: 9000-30-0
EINECS: 232-536-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR