L(-)-Tryptophan ≥99%
Supplier: BeanTown Chemical
Synonyms:
L-α-Amino-3-indolepropionic acid
, (S)-2-Amino-3-(3-indolyl)propionic acid
, H-Trp-OH
, L-Tryptophan
, Tryptophan
215060-100G
215060-25G
215060-500G
BT215060-500GEA
371.85
USD
BT215060-500G
BT215060-100G
BT215060-25G
L(-)-Tryptophan ≥99%
L(-)-Tryptophan
- RTECS: YN6130000
- Powder
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol Melting Pt: 280…285 °C (dec.) |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 EINECS: 200-795-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...