Methylene blue 1% (w/v) in water
Supplier: BeanTown Chemical
Synonyms:
Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
214895-100ML
214895-500ML
BT214895-100MLEA
45.01
USD
BT214895-100ML
BT214895-500ML
Methylene blue 1% (w/v) in water
Methylene blue
CAS: 61-73-4; MDL No: MFCD00012111
Liquid; Molecular Formula: C16H18ClN3S; MW: 319.85
- Liquid
Formula:
C₁₆H₁₈ClN₃S Boiling Pt: 100 °C (212 °F) Melting Pt: 0 °C (32 °F) Density: 1 g/cm³ (8.345 lbs/gal) |
MDL Number:
MFCD00150008 CAS Number: 61-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...