Gold ≥99.95% (trace metals basis), spherical powder APS 1.5-3.0 µm

Supplier: BeanTown Chemical

214890-1G 214890-250MG 214890-5G
BT214890-1GEA 207 USD
BT214890-1G BT214890-250MG BT214890-5G
Gold ≥99.95% (trace metals basis), spherical powder APS 1.5-3.0 µm
Gold

CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436; RTECS: MD5420000 Powder, S.A. 0.10-0.35 m2/g ; Molecular Formula: Au; MW: 196.97 Melting Point: 1063°; Boiling Point: 2808° Density (g/mL): 19.3


  • RTECS: MD5420000
  • Powder, S.A. 0.10-0.35 m²/g

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/mL
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR