Silica gel, Pore size: 60 Å ≥99.5% (trace metals basis) 0.040-0.063mm; 230-400 mesh

Supplier: BeanTown Chemical

214770-1KG 214770-250G 214770-50G
BT214770-1KGEA 242.43 USD
BT214770-1KG BT214770-250G BT214770-50G
Silica gel, Pore size: 60 Å ≥99.5% (trace metals basis) 0.040-0.063mm; 230-400 mesh
Silica gel

CAS: 63231-67-4; EC No: 231-545-4; MDL No: MFCD00011232; RTECS: VV7565000 Powder, S.A. 500-600 m2/g Melting Point: 1600°; Boiling Point: 2230° Hygroscopic


  • RTECS: VV7565000
  • Hygroscopic
  • Powder, S.A. 500-600 m²/g

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: >1600 °C
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR