Magnesium chloride, anhydrous 99

Supplier: BeanTown Chemical

214515-1KG 214515-5KG
BT214515-1KGEA 48.25 USD
BT214515-1KG BT214515-5KG
Magnesium chloride, anhydrous 99
Magnesium chloride

CAS: 7786-30-3; EC No: 232-094-6; MDL No: MFCD00011106; RTECS: OM2800000 Crystalline; Linear Formula: MgCl2; MW: 95.22 Melting Point: 714° Density (g/mL): 2.32 Hygroscopic


  • RTECS: OM2800000
  • Hygroscopic
  • Crystalline

Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412
Melting Pt: 715
Density: 2.32 g/mL
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR