Sodium chloride ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Saline, NaCl, Sea salt, Halite

214380-10KG 214380-2KG 214380-500G
BT214380-10KGEA 162.96 USD
BT214380-10KG BT214380-2KG BT214380-500G
Sodium chloride ≥99.0% ACS
Sodium chloride

CAS: 7647-14-5; EC No: 231-598-3; MDL No: MFCD00003477; RTECS: VZ4725000 Powder; Linear Formula: NaCl; MW: 58.44 Melting Point: 801°; Boiling Point: 1413° Density (g/mL): 2.165; Refractive Index: 1.544


  • RTECS: VZ4725000
  • Refractive index: 1.544
  • Powder

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.165 g/mL
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay 99.0% min.
Barium (Ba) Passes test
Bromine (Br) ≤0.01%
Calcium (Ca) ≤0.002%
Chlorate and nitrate (as NO3) ≤0.003%
Iron (Fe) ≤2ppm
Iodine (I) ≤0.002%
Potassium (K) ≤0.005%
Magnesium (Mg) ≤0.001%
Phosphate (PO4) ≤5ppm
Sulfate (SO4) ≤0.004%
Insoluble material ≤0.005%
Trace metals (as Pb) ≤5ppm
pH, 5% solution at 25°C 5.0-9.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR