Magnesium oxide ≥99.95%
Supplier: BeanTown Chemical
Synonyms:
Magnesia usta
214205-100G
214205-25G
BT214205-100GEA
222
USD
BT214205-100G
BT214205-25G
Magnesium oxide ≥99.95%
Magnesium oxide
CAS: 1309-48-4; EC No: 215-171-9; MDL No: MFCD00011109
3-12mm Fused Lump; Molecular Formula: MgO ; MW: 40.30
Melting Point: 2852°; Boiling Point: 3600°
Density (g/mL): 3.58; Refractive Index: 1.736
Air Sensitive, Moisture Sensitive
- Refractive index: 1.736
- Air Sensitive, Moisture Sensitive
- Fused lump
Formula:
MgO Boiling Pt: 3600 °C Melting Pt: 2852 °C Density: 3.58 g/mL |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...