Copper ≥99.995% (trace metals basis), foil, Thickness 0.5 mm
Supplier: BeanTown Chemical
214015-50X250
214015-50X50MM
BT214015-50X25EA
345
USD
BT214015-50X25
BT214015-50X50
Copper ≥99.995% (trace metals basis), foil, Thickness 0.5 mm
Copper
CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965; RTECS: GL5325000
Solid; Molecular Formula: Cu; MW: 63.55
Melting Point: 1083.4°; Boiling Point: 2567°
Density (g/mL): 8.94
- RTECS: GL5325000
- Solid
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/mL |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...