Calcium Carbonate, Chelometric Standard, ACS, 99.95 to 100.05%

Supplier: BeanTown Chemical

213950-100G 213950-500G
BT213950-100GEA 70.39 USD
BT213950-100G BT213950-500G
Calcium Carbonate, Chelometric Standard, ACS, 99.95 to 100.05%
Standards Cationic Standards Calcium Standards

May be used as a chelometric standard for the standardization of EDTA solution.


  • Density: 2.93
  • EC Number: 207-439-9
  • Merck Number: 14,1657

Order Now

SPECIFICATIONS

Environmentally Preferable
CAS No. 471-34-1
MDL No. MFCD00010906
Molecular Formula CaCO₃

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR