Magnesium sulfate, anhydrous ≥99.5%

Supplier: BeanTown Chemical

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

213630-10KG 213630-2KG 213630-500G
BT213630-10KGEA 540.27 USD
BT213630-10KG BT213630-2KG BT213630-500G
Magnesium sulfate, anhydrous ≥99.5%
Magnesium sulphate

CAS: 7487-88-9; EC No: 231-298-2; MDL No: MFCD00011110; RTECS: OM4500000 Powder; Linear Formula: MgSO4; MW: 120.37 Melting Point: 1124° (decomposes) Density (g/mL): 1.07 Hygroscopic


  • RTECS: OM4500000
  • Hygroscopic
  • Powder

Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 (dec.)
Density: 1.07 g/mL
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR