Polyethylene glycol 600

Supplier: BeanTown Chemical

Synonyms: Ethylene glycol polymer average Mn 600, PEG 600

213500-1KG 213500-250G
BT213500-1KGEA 92.31 USD
BT213500-1KG BT213500-250G
Polyethylene glycol 600
Polyethylene glycol 600

CAS: 25322-68-3; EC No: 500-038-2; MDL No: MFCD00081839; RTECS: TQ3800000 Flakes; Linear Formula: H(OCH2CH2)nOH; MW ≈600 Melting Point: 20-25°; Flash point: 252°C (485°F) Density (g/mL): 1.128; Refractive Index: 1.467


  • RTECS: TQ3800000
  • Refractive index: 1.467
  • Flakes

Formula: H(OCH₂CH₂)nOH
MW: 600 g/mol
Melting Pt: 20…25 °C
Density: 1.128 g/mL
Flash Pt: 252 °C (485 °F)
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR