Cobalt ≥99.9% (trace metals basis), foil 50x50 mm, Thickness 0.127 mm
Supplier: BeanTown Chemical
213475-1PC
213475-5PC
BT213475-1PCEA
161
USD
BT213475-1PC
BT213475-5PC
Cobalt ≥99.9% (trace metals basis), foil 50x50 mm, Thickness 0.127 mm
Cobalt
CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935
Solid; Molecular Formula: Co; MW: 58.93
Melting Point: 1495°; Boiling Point: 2900°
Density (g/mL): 8.92
- Solid
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 Melting Pt: 1495 Density: 8.92 g/mL |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...