Iron(II,III) oxide 97

Supplier: BeanTown Chemical

Synonyms: Magnetite, Triiron tetraoxide, Iron(II,III) oxide

213435-1KG 213435-5KG
BT213435-1KGEA 57.34 USD
BT213435-1KG BT213435-5KG
Iron(II,III) oxide 97
Iron (II,III) oxide

CAS: 1317-61-9; EC No: 215-277-5; MDL No: MFCD00011010 -325 Mesh Powder; Molecular Formula: Fe3O4; MW: 231.54 Melting Point: 1538° Density (g/mL): 5.2


  • -325 Mesh Powder

Formula: Fe₃O₄
MW: 231.54 g/mol
Melting Pt: 1560
Density: 5.2 g/mL
MDL Number: MFCD00011010
CAS Number: 1317-61-9
EINECS: 215-277-5
Merck Index: 12,04087

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR