Ethyl acetoacetate ≥99%

Supplier: BeanTown Chemical

Synonyms: Acetoacetic acid ethyl ester, EAA

213425-2.5KG 213425-500G
BT213425-2.5KGEA 82.6 USD
BT213425-2.5KG BT213425-500G
Ethyl acetoacetate ≥99%
Ethyl acetoacetate

CAS: 141-97-9; EC No: 205-516-1; MDL No: MFCD00009199; RTECS: AK5250000 Liquid; Linear Formula: CH3COCH2COOCH2CH3; Molecular Formula: C6H10O3; MW: 130.14 Melting Point: -43°; Boiling Point: 181°; Flash point: 73°C (163°F) Density (g/mL): 1.029


  • RTECS: AK5250000
  • Liquid

Formula: C₆H₁₀O₃
Boiling Pt: 181 °C
Melting Pt: –43 °C
Density: 1.029 g/mL
Flash Pt: 73 °C (163 °F)
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR