(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-tert-Butyl 2-Methyl (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-Butyl 2-Methyl Ester, N-Boc-cis-4-hydroxy-L-proline Methyl Ester, Boc-cis-Hyp-OMe

213085-1G 213085-250MG 213085-5G
BT213085-1GEA 171 USD
BT213085-1G BT213085-250MG BT213085-5G
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥97%
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate

CAS: 102195-79-9; MDL No: MFCD00237541 Solid; Molecular Formula: C11H19NO5; MW: 245.27 Melting Point: 82-86°


  • Solid

Formula: C₁₁H₁₉NO₅
MW: 245.28 g/mol
Melting Pt: 82…86 °C
MDL Number: MFCD00237541
CAS Number: 102195-79-9

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR