(Bis(trifluoroacetoxy)iodo)benzene ≥97%
Supplier: BeanTown Chemical
212900-25G
212900-5G
BT212900-25GEA
104.32
USD
BT212900-25G
BT212900-5G
(Bis(trifluoroacetoxy)iodo)benzene ≥97%
(Bis(trifluoroacetoxy)iodo)benzene
CAS: 2712-78-9; EC No: 221-808-1; MDL No: MFCD00009672; RTECS: DA3525000
Crystalline; Molecular Formula: C10H5F6IO4 ; MW: 430.04
Melting Point: 121-125°
Light Sensitive, Moisture Sensitive
- RTECS: DA3525000
- Light Sensitive, Moisture Sensitive
- Crystalline
Formula:
C₁₀H₅F₆IO₄ Melting Pt: 121…125 °C |
MDL Number:
MFCD00009672 CAS Number: 2712-78-9 EINECS: 221-808-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...