Sodium chlorodifluoroacetate ≥97%
Supplier: BeanTown Chemical
212825-100G
212825-25G
212825-500G
BT212825-100GEA
232
USD
BT212825-100G
BT212825-25G
BT212825-500G
Sodium chlorodifluoroacetate ≥97%
Sodium chlorodifluoroacetate
CAS: 1895-39-2; EC No: 217-586-0; MDL No: MFCD00064771
Powder; Linear Formula: ClCF2COONa; MW: 152.46
Melting Point: 196-198°
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₂ClF₂NaO₂ Melting Pt: 196…198 °C |
MDL Number:
MFCD00064771 CAS Number: 1895-39-2 EINECS: 217-586-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...