Isochroman ≥99%
Supplier: BeanTown Chemical
Synonyms:
3,4-Dihydro-1H-2-benzopyran
212655-10G
212655-50G
BT212655-10GEA
40
USD
BT212655-10G
BT212655-50G
Isochroman ≥99%
Isochroman
CAS: 493-05-0; EC No: 207-774-0; MDL No: MFCD00006913
Liquid; Molecular Formula: C9H10O ; MW: 134.18
Flash point: 66°C (150°F)
Density (g/mL): 1.067; Refractive Index: 1.545
- Refractive index: 1.545
- Liquid
Formula:
C₉H₁₀O Density: 1.067 g/mL Flash Pt: 66 °C (150 °F) |
MDL Number:
MFCD00006913 CAS Number: 493-05-0 EINECS: 207-774-0 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...