9-Ethylcarbazole ≥99%

Supplier: BeanTown Chemical

212595-1KG 212595-250G 212595-50G
BT212595-1KGEA 218 USD
BT212595-1KG BT212595-250G BT212595-50G
9-Ethylcarbazole ≥99%
9-Ethylcarbazole

CAS: 86-28-2; EC No: 201-660-4; MDL No: MFCD00004967; RTECS: FE6225700 Flakes; Molecular Formula: C14H13N ; MW: 195.26 Melting Point: 68-70°; Boiling Point: 348°; Flash point: 186°C (367°F)


  • RTECS: FE6225700
  • Flakes

Formula: C₁₄H₁₃N
Boiling Pt: 348 °C
Melting Pt: 68…70 °C
Flash Pt: 186 °C (367 °F)
MDL Number: MFCD00004967
CAS Number: 86-28-2
EINECS: 201-660-4

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR