1-Bromotetradecane ≥98%
Supplier: BeanTown Chemical
Synonyms:
Myristyl bromide
212575-100G
212575-500G
212575-2.5KG
BT212575-100GEA
32.44
USD
BT212575-100G
BT212575-2.5KG
BT212575-500G
1-Bromotetradecane ≥98%
1-Bromotetradecane
- Refractive index: 1.460
- Liquid
Formula:
BrH₂C(CH₂)₁₂CH₃ MW: 277.29 g/mol Boiling Pt: 175…178 °C/20 mmHg Melting Pt: 5…6 °C Density: 0.932 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00000228 CAS Number: 112-71-0 EINECS: 203-999-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...