Potassium hydrogen carbonate 99.7-100.5% ACS

Supplier: BeanTown Chemical

Synonyms: Potassium bicarbonate

212200-100G 212200-500G
BT212200-100GEA 23.98 USD
BT212200-100G BT212200-500G
Potassium hydrogen carbonate 99.7-100.5% ACS
Potassium hydrogen carbonate

CAS: 298-14-6; EC No: 206-059-0; MDL No: MFCD00011402 Granular; Linear Formula: KHCO3; MW: 100.12 Density (g/mL): 2.17 Hygroscopic


  • Hygroscopic
  • Granular

Formula: KHCO₃
Density: 2.17 g/mL
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0

Order Now

Specification Test Results

Assay 99.7-100.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR