Manganese(II) acetate tetrahydrate
Supplier: BeanTown Chemical
211710-100G
211710-2KG
211710-500G
BT211710-100GEA
26.46
USD
BT211710-100G
BT211710-2KG
BT211710-500G
Manganese(II) acetate tetrahydrate
Manganese (II) acetate tetrahydrate
CAS: 6156-78-1; EC No: 211-334-3; MDL No: MFCD00062552; RTECS: AI5775000
Crystalline; Linear Formula: (CH3COO)2Mn·4H2O; Molecular Formula: C4H6MnO4·4H2O; MW: 245.09
Melting Point:
- RTECS: AI5775000
- Crystalline
Formula:
(H₃CCOO)₂Mn·4H₂O Melting Pt: >300 °C Density: 1.589 g/mL |
MDL Number:
MFCD00062552 CAS Number: 6156-78-1 EINECS: 211-334-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...