4-Chloro-3-iodophenol ≥98%

Supplier: BeanTown Chemical

211420-1G 211420-5G
BT211420-5GEA 120.47 USD
BT211420-5G BT211420-1G
4-Chloro-3-iodophenol ≥98%
4-Chloro-3-iodophenol

  • Light Sensitive
  • Solid

Formula: C₆H₄ClIO
MW: 254.45 g/mol
Melting Pt: 73…79 °C
MDL Number: MFCD00070769
CAS Number: 202982-72-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR