Barium carbonate, Technical Grade

Supplier: BeanTown Chemical

145930-100G 145930-2KG 145930-500G
BT145930-100GEA 24.23 USD
BT145930-100G BT145930-2KG BT145930-500G
Barium carbonate, Technical Grade
Barium carbonate

CAS: 513-77-9; EC No: 208-167-3; MDL No: MFCD00003448; RTECS: CQ8600000 Powder; Linear Formula: BaCO3; MW: 197.35


  • RTECS: CQ8600000
  • Powder

Formula: BaCO₃
MW: 197.34 g/mol
Melting Pt: 1350
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1 III
Merck Index: 13,00972

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR