Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS), 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol, 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
145785-100G
145785-25G
145785-500G
BT145785-100GEA
101.1
USD
BT145785-100G
BT145785-25G
BT145785-500G
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥98%
Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)
CAS: 6976-37-0; EC No: 230-237-7; MDL No: MFCD00002853
Crystalline/Powder; Molecular Formula: C8H19NO5 ; MW: 209.24
- Crystalline/Powder
Formula:
C₈H₁₉NO₅ MW: 209.24 g/mol |
MDL Number:
MFCD00002853 CAS Number: 6976-37-0 EINECS: 230-237-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...