Diyttrium trioxide ≥99.9%, pellets

Supplier: BeanTown Chemical

Synonyms: Yttrium oxide, Yttrium (III) oxide

145770-100G 145770-500G
BT145770-100GEA 503 USD
BT145770-100G BT145770-500G
Diyttrium trioxide ≥99.9%, pellets
Diyttrium trioxide

CAS: 1314-36-9; EC No: 215-233-5; MDL No: MFCD00011473; RTECS: ZG3850000 Pellets, approx. 10mm dia x 5mm thick; Linear Formula: Y2O3; MW: 225.81 Melting Point: 2410°; Boiling Point: 4300° Density (g/mL): 5.01 Air Sensitive


  • RTECS: ZG3850000
  • Air Sensitive
  • Pellets, approx. 10mm dia x 5mm thick

Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300
Melting Pt: 2410
Density: 5.01 g/mL
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR