4-Dodecylbenzenesulfonyl azide, soft type

Supplier: BeanTown Chemical
Warning

Synonyms: N-diazo-2-dodecylbenzenesulfonamide, N-diazo-2-dodecylbenzenesulphonamide

145620-100G 145620-25G
BT145620-100GEA 203 USD
BT145620-100G BT145620-25G
4-Dodecylbenzenesulfonyl azide, soft type
4-Dodecylbenzenesulfonyl azide

CAS: 79791-38-1; MDL No: MFCD00274161 Liquid; Molecular Formula: C18H29N3O2S; MW: 351.51 Boiling Point: 140° (decomposes); Flash point: 110°C (230°F) Density (g/mL): 1.051; Refractive Index: 1.510 Light Sensitive


  • Refractive Index: 1.510
  • Light Sensitive

Formula: C₁₈H₂₉N₃O₂S
MW: 351.51 g/mol
Boiling Pt: 140 °C (dec.)
Density: 1.051 (g/mL)
Flash Pt: >110 °C
MDL Number: MFCD00274161
CAS Number: 79791-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR