Silver ≥99.9% (trace metals basis), wire Ø 0.25 mm, annealed
Supplier: BeanTown Chemical
145500-100M
145500-25M
145500-500M
BT145500-100MEA
606
USD
BT145500-100M
BT145500-25M
BT145500-500M
Silver ≥99.9% (trace metals basis), wire Ø 0.25 mm, annealed
Silver
CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000
Solid; Molecular Formula: Ag; MW: 107.87
Melting Point: 960°; Boiling Point: 2212°
Density (g/mL): 10.49
- RTECS: VW3500000
- Solid
Formula:
Ag Boiling Pt: 2212 °C Melting Pt: 960 °C Density: 10.49 g/mL |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...