1-Phenyl-1,3-butandione ≥98%

Supplier: BeanTown Chemical

Synonyms: Benzoylacetone1-Phenylbutane-1,3-dione1-Phenyl-1,3-butanedione1-Benzoylacetone

145435-1KG
BT145435-1KGEA 643.86 USD
BT145435-1KG
1-Phenyl-1,3-butandione ≥98%
1-Phenyl-1,3-butandione

  • RTECS: EK3540200
  • Powder

Formula: C₆H₅C(O)CH₂C(O)CH₃
MW: 162.19 g/mol
Melting Pt: 54…56 °C
Density: 1.09 g/mL
MDL Number: MFCD00008786
CAS Number: 93-91-4
EINECS: 202-286-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR