Tetraethylene glycol ≥99%

Supplier: BeanTown Chemical

Synonyms: 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanolTetraglycol

145420-500ML 145420-2.5L
BT145420-2.5LEA 117.22 USD
BT145420-2.5L BT145420-500ML
Tetraethylene glycol ≥99%
Tetraethylene glycol

  • RTECS: XC2100000
  • Refractive index: 1.459
  • Liquid

Formula: C₈H₁₈O₅
MW: 194.23 g/mol
Boiling Pt: 313…314 °C
Melting Pt: –5.6 °C
Density: 1.125 g/mL
Flash Pt: 193 °C (379 °F)
MDL Number: MFCD00002879
CAS Number: 112-60-7
EINECS: 203-989-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR